Clofibrate

Proveedor: MP Biomedicals

02190342.2 0219034205
ICNA02190342.2EA 31.2 EUR
ICNA02190342.2 ICNA0219034205
Clofibrate
Clofibrate

Practically insoluble in water. Miscible with ethanol, acetone, chloroform, ether.

Formula: C₁₂H₁₅ClO₃
Número MDL: MFCD00000615
Núm. CAS: 637-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR