Isodesmosine ≥99%, yellow solid

Proveedor: MP Biomedicals

Sinónimos: 2-(4-Amino-4-carboxybutyl)-1-(5-amino-5-carboxypentyl)-3,5-bis(3-amino-3-carboxypropyl)pyridinium chloride, 2-amino-6-[3,5-bis(3-amino-4-hydroxy-4-oxobutyl)-2-(4-amino-5-hydroxy-5-oxopentyl)pyridin-1-ium-1-yl]hexanoic acid, Iso-desmosin

0219137901 0219137910
ICNA0219137901EA 160 EUR
ICNA0219137901 ICNA0219137910
Isodesmosine ≥99%, yellow solid
Isodesmosine

Isodesmosine is prepared according to the method of Starcher, et al. from hydrolysed bovine neck ligament by solvent extraction and chromatography on Dowex 50. It is chromatographically purified hygroscopic crystalline powder.


Precaución: Research Use Only (RUO).

Formula: C₂₄H₄₀N₅O₈+
Temperatura de almacenaje: Nevera
Número MDL: MFCD00211039
Núm. CAS: 991-01-5

Order Now

Resultados test especificaciones

Identity Test Passes
Source Bovine Neck Ligament
Purity (amino acid analysis) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR