Diatrizoic acid dihydrate, en polvo

Proveedor: MP Biomedicals

Sinónimos: 3,5-Diacetamido-2,4,6-triiodobenzoic acid dihydrate

0219150110 0219150180
ICNA0219150110EA 58.1 EUR
ICNA0219150110 ICNA0219150180
Diatrizoic acid dihydrate, en polvo
Diatrizoic acid dihydrate

Very slightly soluble in water and in alcohol; soluble in dimethylformamide and in alkali hydroxide solutions.

Formula: C₁₁H₉I₃N₂O₄·2H₂O
MW: 649.95 g/mol
Número MDL: MFCD00069960
Núm. CAS: 50978-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR