Accessories for Rapid Equilbrium Dialysis (RED) Device, Pierce™
Proveedor: Thermo Fisher Scientific
89810 89809 90004 90005
PIER89810EA 2480 EUR
PIER89810 PIER89809 PIER90004 PIER90005
Accessories for Rapid Equilbrium Dialysis (RED) Device, Pierce™
Tubos Tubos para diálisis
These are single-use, disposable inserts made up of two side-by-side chambers separated by a vertical cylinder of dialysis membrane validated for minimal nonspecific binding.
  • Disposable inserts require no presoaking, assembly, or specialized equipment
  • Large dialysis surface area accelerates equilibrium giving rise to short incubation time
  • Ideal molecular-weight (8 kDa) cutoff for protein-drug binding studies
  • Regenerated cellulose with low glycerol content as a humectant
This requires no extensive assembly steps or specialsed equipment, and each chamber or well is easily accessible from the top of the insert after insertion in the base plate. Additionally, the high surface-to-volume ratio of the membrane compartment allows rapid dialysis, where equilibrium can be reached in 4 hours with high levels of reproducibility and accuracy. In many cases, experiments can be completed in less than 100 minutes.