Affinity gel chromatography cartridges, Pierce™ Melon™
Proveedor: Thermo Fisher Scientific
89932 89933
PIER89932EA 308 EUR
PIER89932 PIER89933
Affinity gel chromatography cartridges, Pierce™ Melon™
Columnas de cromatografía
The Melon™ gel chromatography cartridge is a convenient, ready to use prepacked chromatography column for the isolation and purification of antibodies.
  • IgG purification with Melon™ gel resin does not involve binding, washing, and multiple elution steps required by other proteins
  • Antibody purification from serum is four to six times faster than Protein A or G methods
  • For many species of serum, antibody recovery is greater than 90% with purity greater than 80%
  • Reduced need to concentrate antibody before storage or use
  • No harsh antibody elution conditions helps retain IgG activity
  • Melon™ gel resin can be used for multiple antibody purifications
This purification resin offers a simpler and faster alternative to traditional methods for purification of human, mouse and rabbit IgG from serum and other sources. It binds and removes serum proteins, thereby allowing pure IgG to be collected in the flow-through fraction. The resin contains a proprietary ligand that retains most protein found in serum, ascites and culture supernatants, while allowing IgG to pass through the support and be collected.
These cartridges are compatible with major automated liquid-chromatography systems or manual syringe processing, and attach directly to ÄKTA™ or FPLC systems without additional connectors.
Información de suministro: Each column pack includes a purification buffer pack (BupH™, 200 ml of 100X buffer), regenerant BupH™ pack (0,5 L of 1X solution) and an accessory pack with one female Luer-Lok™ adapter, one connector fitting, one column plug and one or two bottom caps.