Affinity chromatography cartridges, streptavidin, Pierce™
Proveedor: Thermo Fisher Scientific
87739 87740
PIER87739EA 303 EUR
PIER87739 PIER87740
Affinity chromatography cartridges, streptavidin, Pierce™
Columnas de cromatografía
Pierce™ high capacity streptavidin chromatography cartridges are convenient, ready to use devices for separating biotinylated molecules from non-biotinylated molecules and for purifying antigens using biotinylated antibodies.
  • Enable fast, easy and reproducible chromatographic separations
  • Can be regenerated multiple times for affinity purification
  • Capacity greater than 10 mg biotinylated BSA/ml of resin
high capacity streptavidin resin has two to three times higher binding capacity than the standard streptavidin agarose resin and is ideal when a high amount of recovery is required. The cartridges are effective for assay development and immunoprecipitation and for physically separating DNA strands produced in a polymerase chain reaction by incorporating biotin in one of the amplification polymers.