Kits, test peróxido de hidrógeno, Spectroquant®, Supelco®
Proveedor: Merck
1.14731.0001 1.18789.0001 1.18789.0001_T
1.14731.0001EA 206 EUR
1.14731.0001 1.18789.0001 1.18789.0001_T
Kits, test peróxido de hidrógeno, Spectroquant®, Supelco®
Kits para test medioambiente
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Kits de prueba reactivos Spectroquant®
-Highly stable, ready to use reagent mixtures in a user friendly design
-Free selection of appropriate cell format according to the measuring range
-Long shelf life up to 3 years - all at room temperature
-Automatic selection of the correct method with help of AutoSelector barcode identification
Note: El rango ofertado en la siguiente tabla hace referencia a los instrumentos Prove y NOVA. Para algunos kits el rango de medida disponible para los instrumentos Spectroquant® Multy y Move 100 puede estar reducido.