Guantes industriales, uvex phynomic lite
Proveedor: UVEX
6004010 6004011 6004006 6004007 6004008 6004009
112-4504EA 55.8 EUR
112-4504 112-4505 112-4327 112-4328 112-4329 112-4340
Guantes industriales, uvex phynomic lite
Guantes
The uvex phynomic lite is the lightest safety glove in its class. The glove liner made of polyamide/elastane features an extremly thin aqua-polymer impregnation on palm and fingertips.
  • Excellent dexterity
  • Excellent dry grip
  • Very high level of breathability
  • Coating is extremely thin but still highly durable
Ideal for precision assembly work and other precision work. Suitable for use in dry and slightly damp areas.
Certificaciones: EN 388:2016 (2121X)
Aprovado para contacto con alimentos
OEKO-TEX® Standard 100