tert-Butyl (3S)-3-amino-5-methylhexanoate 95%

Proveedor: Thermo Scientific
Peligro

337931000 337935000
ACRO337931000EA 66.8 EUR
ACRO337931000 ACRO337935000
tert-Butyl (3S)-3-amino-5-methylhexanoate 95%
tert-Butyl (3S)-3-amino-5-methylhexanoate

Appearance: Clear colorless to yellow Liquid or solid

Formula: C₁₁H₂₃NO₂
Temperatura de almacenaje: Congelador
Número MDL: MFCD01076260
Núm. CAS: 166023-30-9
UN: 3259
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance Clear colorless to yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4350 (20°C, 589 nm)
GC ≥94 %
Specific optical rotation +11° to +15° (20°C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR