Iron(II) phthalocyanine

Proveedor: Thermo Scientific

Sinónimos: Phthalocyanine Iron(II), Iron phthalocyanine

412610050 412610250
ACRO412610050EA 71.4 EUR
ACRO412610050 ACRO412610250
Iron(II) phthalocyanine
Iron(II) phthalocyanine

CAS No.: 132-16-1

Formula: C₃₂H₁₆FeN₈
MW: 568,38 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00015953
Núm. CAS: 132-16-1

Order Now

Resultados test especificaciones

Appearance Dark green to dark purple to black Powder
Infrared spectrum Conforms
Gravimetry ≥80 %
GC ≥99.8 %
Acidity/alkalinity ≤0.0005 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR