Chlorogenic acid predominantly trans (from coffee seeds) 98%
Proveedor: Thermo Fisher Scientific
109240010
109240050
ACRO109240010EA
117
EUR
ACRO109240010
ACRO109240050
Chlorogenic acid predominantly trans (from coffee seeds) 98%
Chlorogenic acid
CAS No.: 327-97-9
Formula:
C₁₆H₁₈O₉ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00003862 Núm. CAS: 327-97-9 |
Resultados test especificaciones
Appearance | White to yellow to light beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...