Glycyrrhetic acid 98+%

Proveedor: Thermo Fisher Scientific

120150050 120150250
ACRO120150050EA 108 EUR
ACRO120150050 ACRO120150250
Glycyrrhetic acid 98+%
Glycyrrhetic acid

Appearance: White to off-white Crystalline powder

Formula: C₃₀H₄₆O₄
Pto. de fusión: 292…295 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003706
Núm. CAS: 471-53-4

Order Now

Resultados test especificaciones

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 4 h)
Sulfated ash ≤0.2 %
Related substances ≤2.0 % (HPLC)
Specific optical rotation +145° to +154° (20°C, 589 nm) (c=1, dioxane) (on dried substance)
Titration with NaOH 98.0 to 102.0 % (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR