N-Carbobenzyloxy-L-alanine 98%

Proveedor: Thermo Scientific

Sinónimos: N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine

169000050 169000250 169001000
ACRO169000050EA 30.6 EUR
ACRO169000050 ACRO169000250 ACRO169001000
N-Carbobenzyloxy-L-alanine 98%
N-Carbobenzyloxy-L-alanine

CAS No.: 1142-20-7

Formula: C₁₁H₁₃NO₄
MW: 223,23 g/mol
Pto. de fusión: 82…84 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00002640
Núm. CAS: 1142-20-7

Order Now

Resultados test especificaciones

Appearance White Powder
Infrared spectrum Conforms
TLC ≥97.5 %
Appearance of solution (5% soln. in MeOH) clear, colorless
Specific optical rotation -14° to -16° (24°C, 589 nm) (c=2, AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR