N-Carbobenzoxy-L-aspartic acid 98%
Proveedor: Thermo Fisher Scientific
|
Atención
|
Sinónimos:
N-Cbz-L-aspartic Acid, N-Carbobenzyloxy-L-aspartic acid, Z-Asp-OH
172050250
172051000
ACRO172050250EA
74.9
EUR
ACRO172050250
ACRO172051000
N-Carbobenzoxy-L-aspartic acid 98%
N-Carbobenzoxy-L-aspartic acid
CAS No.: 1152-61-0
Formula:
C₁₂H₁₃NO₆ MW: 267.24 g/mol Pto. de fusión: 117…119 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00002719 Núm. CAS: 1152-61-0 |
Resultados test especificaciones
Appearance | White Powder |
Infrared spectrum | Conforms |
Specific optical rotation | +8° to +10° (20°C, 589 nm) (c=1, acetic acid) |
Titration with NaOH | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...