7-Aminocephalosporanic acid 95-102%

Proveedor: Thermo Fisher Scientific
Peligro

184410010 184410050
ACRO184410010EA 46.8 EUR
ACRO184410010 ACRO184410050
7-Aminocephalosporanic acid 95-102%
7-Aminocephalosporanic acid

Appearance: Off-white to beige Crystalline powder

Formula: C₁₀H₁₂N₂O₅S
Pto. de fusión: >300 °C
Temperatura de almacenaje: Nevera
Número MDL: MFCD00005177
Núm. CAS: 957-68-6

Order Now

Resultados test especificaciones

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +88° to +100° (20°C, 589 nm) (c=0.5, KH2PO4/NaOH)
Residue after ignition ≤0.1 % (Platinum crucible)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR