TentaGel® MB-Br

Proveedor: Thermo Fisher Scientific

354480010 354480050
ACRO354480010EA 84.1 EUR
ACRO354480010 ACRO354480050
TentaGel® MB-Br
TentaGel® MB-Br

CAS No.:

Temperatura de almacenaje: Nevera
Número MDL: MFCD00217898
Núm. CAS: 73342-17-3

Order Now

Resultados test especificaciones

Appearance White to yellow Powder
Particle size 140 to 170 µm
Loading 0.4 to 0.6 mmol/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR