Testosterone

Proveedor: Thermo Fisher Scientific

164410050 164410100 164410250
ACRO164410050EA 96.1 EUR
ACRO164410050 ACRO164410100 ACRO164410250
Testosterone
Testosterone

  • White to yellowish-white Crystalline powder or crystals

Formula: C₁₉H₂₈O₂
Número MDL: MFCD00003654
Núm. CAS: 58-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR