HPLC columns, ZORBAX® 300 Å Extend-C18
Proveedor: Agilent
763750-902 763973-902 765750-902 765973-902 770995-902 773995-902 820950-932 821125-932 761775-902 761973-902
HEWL763750-902EA 915 EUR
HEWL763750-902 HEWL763973-902 HEWL765750-902 HEWL765973-902 HEWL770995-902 HEWL773995-902 HEWL820950-932 HEWL821125-932 HEWL761775-902 HEWL761973-902
HPLC columns, ZORBAX® 300 Å Extend-C18
Columnas de cromatografía
Rugged, high and low pH separations of polypeptides and peptides from pH 2 to11,5.
  • Different selectivity possible at high and low pH
  • High efficiency and good recovery of hydrophobic peptides at high pH
Ideal for LC/MS with ammonium hydroxide-modified mobile phase.