Syringe filters, ValuPrep™
Proveedor: Cytiva (Formerly Pall Lab)
4114 4118 6054540 6054541
516-7574EA 1720 EUR
516-7574 516-8925 PALL6054540 PALL6054541
Syringe filters, ValuPrep™
Filtros Filtros de jeringas
ValuPrep syringe filters offer the confidence of Pall membranes in economic 1000 packs.
  • Available in 13 and 25 mm sizes
  • Available in 0,2 and 0,45 µm pore sizes, contains Pall PTFE, PVDF, Nylon and wwPTFE membranes
These syringe filters are ideal for routine laboratory HPLC or UHPLC analyses.