GC columns, silica gel
Proveedor: Agilent
G3591-80108 G3591-80141 G3591-80142
HEWLG3591-80108EA 296 EUR
HEWLG3591-80108 HEWLG3591-80141 HEWLG3591-80142
GC columns, silica gel
Columnas de cromatografía
Packed GC columns are designed and manufactured to offer excellent and reproducible performance for all sample types associated with packed column separations, most important in the hydrocarbon processing industry.
  • Highly efficient and rigorous packing technology assures column-to-column reproducibility and ultimate efficiency
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance