L(-)-Glucosa, polvo cristalino
Proveedor: MP Biomedicals
Sinónimos:
(2S,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal
L-Glucose was once proposed as a low-calorie sweetener and it is suitable for patients with diabetes mellitus, but it was never marketed due to excessive manufacturing costs. The acetate derivative of L-glucose, L-glucose pentaacetate, was found to stimulate insulin release, and might therefore be of therapeutic value for type 2 diabetes.
L-Glucose was also found to be a laxative, and has been proposed as a colon-cleansing agent which would not produce the disruption of fluid and electrolyte levels associated with the significant liquid quantities of bad-tasting osmotic laxatives conventionally used in preparation for colonoscopy.
Soluble in water (clear, colorless solution).
Precaución: Research Use Only (RUO).
Formula:
C₆H₁₂O₆ MW: 180,16 g/mol |
Número MDL:
MFCD00148913 Núm. CAS: 921-60-8 |
Resultados test especificaciones
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 144 - 155 °C |
Loss on Drying | <0.8% |
Optical Rotation (c=4, H2O, NH4OH) | –50° to –60° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...