Dihydrostreptomycin sesquisulphate, en polvo

Proveedor: MP Biomedicals

Sinónimos: Dihydrostreptomycin sulphate, Dihydrostreptomycin sulfate

0210040505 0210040525 0210040580
ICNA0210040505EA 51.5 EUR
ICNA0210040505 ICNA0210040525 ICNA0210040580
Dihydrostreptomycin sesquisulphate, en polvo
Dihydrostreptomycin sesquisulphate

Dihydrostreptomycin sulphate is an aminoglycoside antibiotic. It inhibits protein synthesis by interfering with translation.


Dihydrostreptomycin sulphate is an antibiotic used in aminoglycoside uptake studies. It has also been used as an electrode for a cochlear amplifier in the hair-cell bundle of lizards.


Mode of action: Inhibits bacterial protein synthesis at the level of the 30S ribosomal subunit and the 16S rRNA.


Add the appropriate amount of powder to water while mixing (not the reverse), as adding water to the powders may result in insoluble mixtures that cannot be filtered without losing potency. If desired, the pH may be adjusted, but a basal media with greater than 2 g/L sodium bicarbonate can easily absorb up to 2% of an acidic antibiotic solution; therefore, an antibiotic solution with an acidic pH would not influence the pH of the media. After ensuring that the resultant solution is transparent, sterile filter and store in small aliquots at appropriate temperature until use. Prior to use of the media, aseptically add the appropriate amount of antibiotic to the basal media in order to obtain the required working concentration. Mix well and sterile filter if asepsis is believed to have been compromised.

Formula: C₂₁H₄₁N₇O₁₂·1.5H₂SO₄
MW: 1461.42 g/mol
Pto. de fusión: 220…235 °C (desc.)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00070252
Núm. CAS: 5490-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR