Sodium diatrizoate ≥99%, white crystalline powder

Proveedor: MP Biomedicals
Peligro

Sinónimos: Diatrizoate sodium, 3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid sodium salt, 3,5-Diacetamido-2,4,6-triiodobenzoic acid sodium salt, Diatrizoic acid sodium salt, Amidotrizoate sodium salt, Sodium 3,5-diacetamido-2,4,6-triiodobenzoate

0210290810 0210290850 0210290880
ICNA0210290810EA 49.5 EUR
ICNA0210290810 ICNA0210290850 ICNA0210290880
Sodium diatrizoate ≥99%, white crystalline powder
Sodium diatrizoate

Sodium Diatrizoate has been shown to cause partial redistribution of tight-junction-associated membrane proteins into a cytoplasmic compartment in cultured monolayer MDCK cells.


Sodium diatrizoate is used as a density gradient reagent in blood cell separation. It has also been utilized as a radiological contrast agent.


Soluble in water (350 mg/mL).


Precaución: Research Use Only (RUO).

Formula: C₁₁H₈I₃N₂NaO₄
MW: 635.898 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00051544
Núm. CAS: 737-31-5

Order Now

Resultados test especificaciones

Form White Powder
Storage Store at Room Temperature (15 - 30 °C). Protect from light.
Loss on Drying 6,0 - 8,0%
pH 7,0 - 9,7 (10% solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR