Amonio cloruro, en polvo
Proveedor: MP Biomedicals
Sinónimos:
Ammonium muriate, Salmiac
0215010980
0215010991
ICNA0215010980EA
25.7
EUR
ICNA0215010980
ICNA0215010991
Amonio cloruro, en polvo
Amonio cloruro
Soluble in water (100 mg/ml - clear, colorless solution). The presence of hydrochloric acid or sodium chloride decrease water solubility. Also soluble in methanol and ethanol.
Soluble in water (100 mg/ml - clear, colorless solution). The presence of hydrochloric acid or sodium chloride decrease water solubility. Also soluble in methanol and ethanol.
Formula:
NH₄Cl |
Número MDL:
MFCD00011420 Núm. CAS: 12125-02-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...