Cefadroxil, white powder

Proveedor: MP Biomedicals
Atención

Sinónimos: Cefadroxil monohydrate, (6R,7R)-7-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

0215058101 0215058105
ICNA0215058101EA 77.5 EUR
ICNA0215058101 ICNA0215058105
Cefadroxil, white powder
Cefadroxil

Antibiotic. Cefadroxil was used to investigate its interaction with hPEPT1 and dipeptide Gly-Sar on basis of steady-state and presteady-state analysis thus testing the hypothesis that variations in drug response among individuals are caused by alterations in the genes involved in drug absorption.


Slightly soluble in water; practically insoluble in alcohol, chloroform, ether.


Precaución: Research Use Only (RUO). Protect from light.

Formula: C₁₆H₁₇N₃O₅S
MW: 363,388 g/mol (anhy)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00865091
Núm. CAS: 50370-12-2

Order Now

Resultados test especificaciones

Identity Test Passes
Potency Report Result (µg/mg)
Melting Point 190 - 205 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR