2'-Deoxyadenosine-5'-diphosphate trisodium salt ≥97%, white powder
Proveedor: MP Biomedicals
|
Peligro
|
0215079410
0215079425
0215079480
ICNA0215079410EA
70.8
EUR
ICNA0215079410
ICNA0215079425
ICNA0215079480
2'-Deoxyadenosine-5'-diphosphate trisodium salt ≥97%, white powder
2'-Deoxyadenosine-5'-diphosphate trisodium salt
2'-Deoxyadenosine 5'-diphosphate (dADP) may be used for the synthesis of deoxyadenylate oligonucleotides with enzymes such as polynucleotide phosphorylase from Escherichia coli.
dADP is used as an inhibitor of bacterial poly(A) polymerase. DeoxyADP may be use as an alternative substrate or inhibitor for studies of ATPase and DNA and RNA polymerase specificity.
Precaución: Research Use Only (RUO).
Formula:
C₁₀H₁₃N₅Na₂O₉P₂ Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00083610 Núm. CAS: 72003-83-9 |
Resultados test especificaciones
Identity Test | Passes |
Purity | ≥97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...