2-Deoxy-D-galactose, white powder

Proveedor: MP Biomedicals

0215080001 0215080005 0215080025
ICNA0215080001EA 78.1 EUR
ICNA0215080001 ICNA0215080005 ICNA0215080025
2-Deoxy-D-galactose, white powder
2-Deoxy-D-galactose

Inhibitor of fucosylation.


Precaución: Research Use Only (RUO).

Formula: C₆H₁₂O₅
MW: 164,16 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00014649
Núm. CAS: 1949-89-9

Order Now

Resultados test especificaciones

Identity Test Passes
Melting Point 109 - 119°C
FTIR Conforms to Standard
Optical Rotation (c=4, H₂O) +54.8° to +66.3°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR