p-Fenilendiamina, escamas

Proveedor: MP Biomedicals
Peligro

Sinónimos: 1,4-Fenilendiamina

0215183080 0215183083
ICNA0215183080EA 79.6 EUR
ICNA0215183080 ICNA0215183083
p-Fenilendiamina, escamas
p-Fenilendiamina

P-Phenylenediamine is an anti-fade reagent added to the mounting medium in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes.


Soluble in ethyl alcohol (EtOH)(50 mg/ml).


Precaución: Research Use Only (RUO). Protect from light, store under nitrogen.

Formula: C₆H₈N₂
MW: 108,14 g/mol
Número MDL: MFCD00007901
Núm. CAS: 106-50-3
UN: 1673
ADR: 6.1,III

Order Now

Resultados test especificaciones

Identity Test Passes
FTIR Conforms to Standard
Melting Point 138 - 149 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR