Alcohol polivinílico, M.W. ~22000, en polvo

Proveedor: MP Biomedicals

Sinónimos: PVOH, PVA, Mowiol®

0215193883 0215193891
ICNA0215193883EA 127 EUR
ICNA0215193883 ICNA0215193891
Alcohol polivinílico, M.W. ~22000, en polvo
Alcohol polivinílico

Polyvinyl alcohol is the raw material to make polymers. Polyvinyl alcohol is used as a emulsion polymerization aid, as protective colloid, to make polyvinyl acetate dispersions.


  • Viscosity (4% aq soln) 4 to 6 cps


Precaución: Research Use Only (RUO).

Formula: [-CH₂CH(OH)-]n
Número MDL: MFCD00081922
Núm. CAS: 9002-89-5

Order Now

Resultados test especificaciones

Identity Test Passes
Viscosity (4% aq soln) 4 - 6 cps


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR