Universol™-es liquid scintillation cocktail, MP Biomedicals
Proveedor: MP Biomedicals
0188248001 0188248004
ICNA0188248001EA 303 EUR
ICNA0188248001 ICNA0188248004
Universol™-es liquid scintillation cocktail, MP Biomedicals
Líquido centelleo
UniverSol™ is a very efficient environmentally safe, liquid scintillation cocktail of the 'gelling' variety. The physical characteristics of UniverSol™-ES are identical to traditional 'TRITON™ and Toluene' type commercial cocktails.
  • Environmentally safe
  • Non toxic and non flammable
  • Multi-use for aqueous samples
  • High tritium efficiency
  • Both clear and translucent gel phases