Benztropine mesylate, white powder

Proveedor: MP Biomedicals

Sinónimos: Cogentin

0215362625 0215362680
ICNA0215362625EA 55.6 EUR
ICNA0215362625 ICNA0215362680
Benztropine mesylate, white powder
Benztropine mesylate

Soluble in water.

Formula: C₂₂H₂₉NO₄S
MW: 403,537 g/mol
Número MDL: MFCD00074784
Núm. CAS: 132-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR