Colofonia, chunks
Proveedor: MP Biomedicals
Sinónimos:
Rosin, gum, Rosin, Gum rosin
0215655790
0215655791
ICNA0215655790EA
82.6
EUR
ICNA0215655790
ICNA0215655791
Colofonia, chunks
Colofonia
Soluble in alcohol (ethanol: 50 mg/ml), benzene, ether. Insoluble in water. Chloroform: soluble 0,1 g/ml, clear, slightly yellow to deep yellow.
Soluble in alcohol (ethanol: 50 mg/ml), benzene, ether. Insoluble in water. Chloroform: soluble 0,1 g/ml, clear, slightly yellow to deep yellow.
Pto. de fusión:
70…130 °C (lit.) Pto. de inflamación: 188 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00132205 Núm. CAS: 8050-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...