Preparative HPLC columns, Ultra Cyano
Proveedor: Restek
9106517 9106518 9106519 9106557 9106558 9106559 9106567 9106568 9106569 9106577 9106578 9106579
REST9106517EA 1640 EUR
REST9106517 REST9106518 REST9106519 REST9106557 REST9106558 REST9106559 REST9106567 REST9106568 REST9106569 REST9106577 REST9106578 REST9106579
Preparative HPLC columns, Ultra Cyano
Columnas de cromatografía
These preparative HPLC columns with spherical shaped traditional monomeric cyanopropylsilane are well suitable for the analysis of protonated bases and also assays where alternate selectivity or confirmation is needed to C18 or C8 column.
  • Used in normal phase, reversed phase and HILIC modes
  • Suited with mobile phase when the pH is between 5 and 7