Solid phase extraction cartridges, Resprep®, C18 / C8
Proveedor: Restek
24051 26030 26031 26034 26036 26037 26038 26039 26040
REST24051EA 251 EUR
REST24051 REST26030 REST26031 REST26034 REST26036 REST26037 REST26038 REST26039 REST26040
Solid phase extraction cartridges, Resprep®, C18 / C8
Columnas de cromatografía
These cartridges are made of glass fiber disks embedded with C18 or C8 bonded silica. These hydrophobic (nonpolar) silica-based adsorbents are ideal to extract hydrophobic analytes from polar matrices, such as water (e.g., pesticides from water).
  • All cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted
  • Extract semivolatile organic compounds
  • Deep-pore design reduces clogging and allows faster flow rates
  • Lower cost than PTFE disks
Certificaciones: Meet requirements for EPA Methods 525.1, 506, 550.1, and 549.1.