Capillary columns, MXT®-1
Proveedor: Restek
MXT®-1
REST70188-109EA 1310 EUR
REST70188-109 REST70177-139 REST70182-283 REST70155-106283 REST70158-109271 REST70122-104271 REST70137-104 REST70137-107 REST70138-273 REST70170-273 REST70158-115283 REST70170-271 REST70158-283 REST70155-106273 REST70188-105 REST70152-097 REST70182-273 REST70155-106271 REST70182-271 REST70185-106273 REST70137-123271 REST70143-283 REST70158-272 REST70158-271 REST70193-115 REST70158-274 REST70158-273 REST70158-277 REST70188-118 REST70193-272 REST70193-273 REST70193-271 REST70185-272 REST70185-271 REST70185-274 REST70185-273 REST70177-273 REST70177-272 REST70142-271 REST70142-273 REST70177-274 REST70177-271 REST70177-107283 REST70177-283 REST70123-273 REST70151-283 REST70170-283 REST70173-105 REST70182-107273 REST70158-105271 REST70155-274 REST70182-107272 REST70155-273 REST70158-105273 REST70155-272 REST70155-271 REST70182-107271 REST70179-101271 REST70193 REST70191 REST70192 REST70155-278 REST70188 REST70189 REST70187 REST70184 REST70152-107283 REST70185 REST70122-274 REST70122-273 REST70122-271 REST70158-105283 REST70193-283 REST70155-283 REST70185-102271 REST70155-280 REST70182 REST70183 REST70181 REST70127-271 REST70127-273 REST70179 REST70152-107 REST70192-112 REST70177 REST70152-104 REST70159-273 REST70173 REST70137-2830 REST70177-107273 REST70125-111273 REST70177-107271 REST70126-283 REST70193-274 REST70150-273 REST70191-104 REST70185-106283 REST70192-271 REST70173-2730 REST70192-273 REST70137-104283 REST70137-283 REST70143-271 REST70109-273 REST70154-283 REST70155-101283 REST70167-104271 REST70126-273 REST70136-2830 REST70182-107283 REST70185-102 REST70185-101 REST70177-085 REST70137-104274 REST70155-2830 REST70185-106 REST70137-104271 REST70137-273 REST70152-107277 REST70152-107273 REST70152-107271 REST70122-283 REST70154-271 REST70173-109271 REST70154-273 REST70137-271 REST70173-105271 REST70173-105273 REST70177-097 REST70150 REST70155-112 REST70144 REST70156-273 REST70142 REST70121-273 REST70143 REST70140 REST70182-123271 REST70141 REST70122-107271 REST70167-107283 REST70101-273 REST70101-274 REST70144-273 REST70173-105283 REST70144-271 REST70191-119 REST70188-105283 REST70137-107283 REST70135 REST70170-102283 REST70188-115400 REST70136 REST70122-107283 REST70139 REST70155-6850M REST70137 REST70138 REST70125-273 REST70185-102283 REST70188-105273 REST70170 REST70136-283 REST70125-271 REST70167 REST70188-283 REST70152-123271 REST70183-271 REST71816 REST70108-276 REST71815 REST70191-271 REST71813 REST71812 REST70191-274 REST71811 REST70109-6850M REST70152-351 REST70136-273 REST70157-109283 REST70153-273 REST70183-273 REST70140-102283 REST70167-119 REST70140-273 REST70140-272 REST70179-271 REST70179-273 REST70101-133 REST70179-272 REST70157 REST70158 REST70155 REST70156 REST70153 REST70154 REST70151 REST70152 REST70117-283 REST70152-123 REST70155-102283 REST70159 REST70120-273 REST70114-283 REST70182-104283 REST70157-283 REST70140-101283 REST70173-276 REST70102 REST70173-277 REST70182-119 REST70151-107283 REST70173-273 REST70170-2730 REST70108 REST70109 REST70106 REST70167-283 REST70107 REST70139-273 REST70105 REST70177-104 REST70177-107 REST70185-283 REST70157-273 REST70157-031 REST70177-104271 REST70170-112273 REST70157-271 REST70177-104273 REST70157-277 REST70152-2730 REST70151-271 REST70182-107 REST70188-271 REST70173-283 REST70188-272 REST70188-273 REST70188-115283 REST70193-115400 REST70182-104 REST70152-065 REST70140-106 REST70137-107271 REST70140-102 REST70124 REST70125 REST70122 REST70123 REST70120 REST70152-2830600 REST70121 REST70152-104271 REST70152-277 REST70128 REST70152-271 REST70129 REST70126 REST70152-273 REST70127 REST70152-274 REST70157-105283 REST70158-103283 REST70188-103283 REST70155-106 REST70129-283 REST70173-271 REST70152-2830 REST70155-102 REST70155-101 REST70137-123283 REST70114 REST70152-104283 REST70111 REST70150-2770 REST70117 REST70152-283 REST70124-273 REST70116
Capillary columns, MXT®-1
Columnas de cromatografía
MXT®-1 columns exhibit long lifetime and very low bleed at high operating temperatures. A proprietary synthesis process eliminates residual catalysts that could cause degradation and increase bleed.
  • Non polar phase, crossbond dimethyl polysiloxane
  • Equivalent to USP G1, G2, G38 phases
  • 114,3 mm standard coil diameter
Ideal for solvent impurities, PCB congeners, gases, natural gas odorants, sulphur compounds, essential oils, hydrocarbons, semivolatiles, pesticides, and oxygenates.