Capillary columns, MXT®-50 (Siltek®-treated stainless steel)
Proveedor: Restek
70552 70555 70558 70570
REST70552EA 573 EUR
REST70552 REST70555 REST70558 REST70570
Capillary columns, MXT®-50 (Siltek®-treated stainless steel)
Columnas de cromatografía
MXT®-50 columns are general purpose columns for pesticides, herbicides, rosin acids, phthalate esters and sterols.
  • Midpolarity phase; Crossbond® diphenyl dimethyl polysiloxane
  • Temperature range up to 300 °C
  • Equivalent to USP G3 phase
Coil diameter: 114,3 mm