Anti-CD28 Mouse Monoclonal Antibody (CF647) [clone: C28/77]

Proveedor: Biotium
BNC470077-500 BNC470077-100
BNC470077-500EA 560 EUR
BNC470077-500 BNC470077-100
Anti-CD28 Mouse Monoclonal Antibody (CF647) [clone: C28/77]
Anticuerpos
Recognizes a glycoprotein of 44-88 kDa, which is identified as CD28. It is the critical T-cell co-stimulatory receptor which provides to the cell the important second activation signal by binding CD80 and CD86 that are expressed by antigen presenting cells. Besides its co-stimulation role, CD28 functions in preventing T-cells from anergic hyporesponsive state or from undergoing premature apoptotic cell death. CD28 is also expressed on human fetal NK cells and some NK cell lines, whereas on murine NK cells the CD28 expression is much broader.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: C28/77
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR