Anti-CMV-p65 Mouse Monoclonal Antibody (Purified) [clone: CMV101]
Proveedor: Biotium
BNUB0101-500 BNUB0101-100
BNUB0101-500EA 511 EUR
BNUB0101-500 BNUB0101-100
Anti-CMV-p65 Mouse Monoclonal Antibody (Purified) [clone: CMV101]
Anticuerpos
Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: CMV101
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: CMV