Anti-S100A1 Mouse Monoclonal Antibody (CF568) [clone: 4C4.9]
Proveedor: Biotium
BNC680143-500 BNC680143-100
BNC680143-500EA 560 EUR
BNC680143-500 BNC680143-100
Anti-S100A1 Mouse Monoclonal Antibody (CF568) [clone: 4C4.9]
Anticuerpos
S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle, Schwann cells, epithelial and myoepithelial cells of the breast, salivary and sweat glands, as well as in glial cells. Neoplasms derived from these cells also express S-100 protein, albeit non-uniformly. A large number of well-differentiated tumors ofMore Product Information