Anti-ITGAL Mouse Monoclonal Antibody (CF647) [clone: Cris-3]

Proveedor: Biotium
BNC470151-500 BNC470151-100
BNC470151-500EA 560 EUR
BNC470151-500 BNC470151-100
Anti-ITGAL Mouse Monoclonal Antibody (CF647) [clone: Cris-3]
Anticuerpos
Recognizes a protein of 180 kDa, identified as CD11a (Leucocyte Workshop II; Code N202). CD11a complex with the 2 subunit of the integrin family, CD18, to form the cell surface heterodimer, LFA-1 or CD11a /C18 (aLbL). LFA-1 is expressed on all leukocytes including lymphocytes, monocytes, and granulocytes. It is involved in leukocyte adhesion to its ligands including intercellular adhesion molecule-1 (ICAM-1 or CD54), ICAM-2 (CD102), ICAM-3 (CD50) and Telencephalin (TLN) and play a role in most immune/inflammatory responses.

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: Cris-3
More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno ITGAL
Environmentally Preferable
Nombre del antígeno CD11a
Sinónimos de antígenos p180|LFA 1 alpha (LFA1A)|ITGAL|Antigen CD11A (p180)|Ly21|Integrin alpha-L|Ly15|lymphocyte function associated antigen 1|alpha polypeptide|Leukocyte Adhesion Glycoprotein LFA1 Alpha Chain
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 647
Clon Clone Cris-3
Reactividad Human
Huésped Mouse
ID del gen 174103
Isotipo IgG2b kappa
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR