Anti-DPP4 Mouse Monoclonal Antibody (Purified) [clone: 134-2C2]

Proveedor: Biotium
BNUB0162-500 BNUB0162-100
BNUB0162-500EA 511 EUR
BNUB0162-500 BNUB0162-100
Anti-DPP4 Mouse Monoclonal Antibody (Purified) [clone: 134-2C2]
Anticuerpos
Recognizes a glycoprotein of 110 kDa, identified as CD26. It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection.

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: 134-2C2
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR