Anti-CD27 Mouse Monoclonal Antibody (CF488A) [clone: 203.6]

Proveedor: Biotium
BNC880163-500 BNC880163-100
BNC880163-500EA 560 EUR
BNC880163-500 BNC880163-100
Anti-CD27 Mouse Monoclonal Antibody (CF488A) [clone: 203.6]
Anticuerpos
Recognizes a protein of a disulfide-linked 120 kDa dimer, identified as CD27 (Workshop VI; Code 6T-028). CD27 is expressed on the majority of peripheral T cells, medullary thymocytes, memory-type B cells, and natural killer cells. It is a transmembrane phosphoglycoprotein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. CD27 binds to its ligand CD70, a member of the TNF family, and induces T-cell co-stimulation and B-cell activation. It also interacts with TRAFs and is involved in activation of NFB and SAPK/JNK and induces apoptosis.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 203.6
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR