Anti-CKB Mouse Monoclonal Antibody (CF568) [clone: 2ba6]

Proveedor: Biotium
BNC680196-500 BNC680196-100
BNC680196-500EA 560 EUR
BNC680196-500 BNC680196-100
Anti-CKB Mouse Monoclonal Antibody (CF568) [clone: 2ba6]
Anticuerpos
Creatine kinases (CK) are a large family of isoenzymes that regulate levels of ATP in subcellular compartments, where they provide ATP at sites of fluctuating energy demand by the transfer of phosphates between creatine and adenine nucleotides. CKs provide the energy of phosphate hydrolysis necessary to drive the normal function of many cellular systems. In cells, the cytosolic CK enzymes consist of two subunits, which can be either B (brain type) or M (muscle type). There are three different isoenzymes: CKMM, CKBB and CKMB. This MAb recognizes the CKBB isoenzyme and does not react with the B subunit in CKMB. It shows minimal reactivity with other human serum proteins

Type: Primary
Antigen: CKB
Clonality: Monoclonal
Clone: 2ba6
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR