Anti-CD4 Mouse Monoclonal Antibody (CF568) [clone: C4/206]

Proveedor: Biotium
BNC680206-500 BNC680206-100
BNC680206-500EA 560 EUR
BNC680206-500 BNC680206-100
Anti-CD4 Mouse Monoclonal Antibody (CF568) [clone: C4/206]
Anticuerpos
Recognizes a protein of 55 kDa, identified as CD4. CD4 is a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: C4/206
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR