Anti-MVP Mouse Monoclonal Antibody (CF594) [clone: 1014]

Proveedor: Biotium
BNC940224-500 BNC940224-100
BNC940224-500EA 560 EUR
BNC940224-500 BNC940224-100
Anti-MVP Mouse Monoclonal Antibody (CF594) [clone: 1014]
Anticuerpos
Recognizes a protein of 104 kDa-110 kDa, characterized as major vault protein (MVP). Vaults are large ribonucleoprotein particles (RNPs) present in all eukaryotic cells. They have a complex morphology, including several small molecules of RNA, but a single protein species. The MVP accounts for >70% of their mass. Their shape is reminiscent of the nucleopore central plug. Treatment of cells with estradiol increases the amount of MVP in nuclear extract. The hormone-dependent interaction of vaults with ER is prevented in vitro by sodium molybdate. Antibodies to estrogen, progesterone and glucocorticoid receptors are able to co-immunoprecipitate the MVP. MVP is overexpressed in many neoplastic tissues and cell lines. Expression of MVP predicts a poor response to chemotherapy.

Type: Primary
Antigen: MVP
Clonality: Monoclonal
Clone: 1014
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR