Anti-Testosterone Mouse Monoclonal Antibody (CF555) [clone: 4E1G2]

Proveedor: Biotium
BNC550250-500 BNC550250-100
BNC550250-500EA 560 EUR
BNC550250-500 BNC550250-100
Anti-Testosterone Mouse Monoclonal Antibody (CF555) [clone: 4E1G2]
Anticuerpos
This MAb is highly specific to testosterone. Its affinity constant for testosterone is ~1010M-1. In competitive binding immunoassay, it reacts with testosterone 100%, 11-beta-hydroxy testosterone 3.3%, 17-alpha-methyl testosterone

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: 4E1G2
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR