Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: MYC275]

Proveedor: Biotium
BNUB0275-500 BNUB0275-100
BNUB0275-500EA 511 EUR
BNUB0275-500 BNUB0275-100
Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: MYC275]
Anticuerpos
The c-Myc protein is a transcription factor, which is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression. The phosphorylation of c-Myc has been investigated and previous studies have suggested a functional association between phosphorylation at Thr58/Ser62 by glycogen synthase kinase 3, cyclin dependent kinase, ERK2 and C-Jun N terminal Kinase (JNK) in cell proliferation and cell cycle regulation. Studies also have shown that c-Myc is essential for tumor cell development in vasculogenesis and angiogenesis that distribute blood throughout the cells, and which brought extensive attention in the development of new therapeutic approach for cancer treatment.

Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: MYC275
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR