Anti-CD48 Mouse Monoclonal Antibody (Biotin) [clone: 156-4H9]

Proveedor: Biotium
BNCB0307-500 BNCB0307-100
BNCB0307-500EA 560 EUR
BNCB0307-500 BNCB0307-100
Anti-CD48 Mouse Monoclonal Antibody (Biotin) [clone: 156-4H9]
Anticuerpos
Reacts with human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of marker for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 156-4H9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR