Anti-Lewis A Mouse Monoclonal Antibody (CF647) [clone: 7LE]
Proveedor: Biotium
BNC470311-500 BNC470311-100
BNC470311-500EA 560 EUR
BNC470311-500 BNC470311-100
Anti-Lewis A Mouse Monoclonal Antibody (CF647) [clone: 7LE]
Anticuerpos
Recognizes a carbohydrate determinant of Gal 1-3(Fuc 1-4) GlcNAc which is blood group antigen Lewis A. It is present primarily on epithelial cells such as colon and kidneys. In the tumors and dedifferentiated tissues, decrease of Lewis A antigen was observed. Lewis A (type 1 chain) is expressed in colonic epithelial cells and may be useful for detection of gastrointestinal tumors, pancreatic cancer, and colorectal tumors. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected onMore Product Information