Anti-CD4 Mouse Monoclonal Antibody (CF488A) [clone: EDU-2]

Proveedor: Biotium
BNC880345-500 BNC880345-100
BNC880345-500EA 560 EUR
BNC880345-500 BNC880345-100
Anti-CD4 Mouse Monoclonal Antibody (CF488A) [clone: EDU-2]
Anticuerpos
Recognizes a protein of 55 kDa, identified as CD4. It is a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.This MAb was characterized as human CD4 antibody at II and IV International Workshop on Human Leukocyte Differentiation Antigens.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: EDU-2
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Chimpanzee, Human, Macaca artctoids, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR